Learn More
Hach Company CL111 Electrode Cable: S7 to Banana (1 meter cable)
Catalog No. 501997492 Shop All Hach Company Products
Encompass
The CL111 type connection cable is for screw cap electrodes (S7) to banana instrument sockets. The cable has a 1 meter length.
Catalog No. 50-199-7492 Supplier Hach Company Supplier No. A94L111
The CL111 type connection cable is for screw cap electrodes (S7) to banana instrument sockets. The cable has a 1 meter length.