Learn More
Hach Company CL114 Electrode Cable: S7 to BNC (1 meter cable)
Catalog No. 501997493 Shop All Hach Company Products
Encompass
The CL114 type connection cable is for screw cap electrodes (S7) to BNC instrument sockets. The cable has a 1 meter length.
Catalog No. 50-199-7493 Supplier Hach Company Supplier No. A94L114
The CL114 type connection cable is for screw cap electrodes (S7) to BNC instrument sockets. The cable has a 1 meter length.