Learn More
Hach Company Non-Assembled Tubing Kit for Hach CL17 Colorimetric Chlorine Analyzer
Catalog No. NC3036735 Shop All Hach Company Products
Encompass
The CL17 Non-assembled Tubing Kit (Hach part 5448000) provides replacement tubing and connectors for installation and maintenance of the CL17 colorimetric chlorine analyzer. Supplied unassembled, the kit allows field technicians to route and assemble tubing to match site requirements.
  • Supplied non-assembled for flexible field installation.
  • Includes tubing and connectors required for fluid connections.
  • Used to replace worn or damaged tubing components.
  • Allows customized tubing runs to fit system layout.
  • Suitable for routine maintenance and repair tasks.
Catalog No. NC3036735 Supplier Hach Company Supplier No. 5448000
The CL17 Non-assembled Tubing Kit (Hach part 5448000) provides replacement tubing and connectors for installation and maintenance of the CL17 colorimetric chlorine analyzer. Supplied unassembled, the kit allows field technicians to route and assemble tubing to match site requirements.
  • Supplied non-assembled for flexible field installation.
  • Includes tubing and connectors required for fluid connections.
  • Used to replace worn or damaged tubing components.
  • Allows customized tubing runs to fit system layout.
  • Suitable for routine maintenance and repair tasks.