Learn More
Hach Company CL17 Pressurized Bottle-Cap Kit for Hach CL17 Chlorine Analyzer
Catalog No. NC3717559 Shop All Hach Company Products
Encompass
The CL17 Pressurized Bottle-Cap Kit (MPN 5449900) from Hach is a bottle-cap pressurization accessory designed for manual calibration and verification of the CL17 Chlorine Analyzer. It enables controlled pressurization of reagent bottles to perform accurate calibration and verification procedures.
  • Enables controlled pressurization of reagent bottles for manual calibration and verification.
  • Includes a luer lock syringe, a one-way check valve, and all necessary luer tubing and cap.
  • Designed for use with chlorine analyzers in field or laboratory calibration workflows.
  • Provides quick installation with the included fittings for efficient maintenance.
Catalog No. NC3717559 Supplier Hach Company Supplier No. 5449900
The CL17 Pressurized Bottle-Cap Kit (MPN 5449900) from Hach is a bottle-cap pressurization accessory designed for manual calibration and verification of the CL17 Chlorine Analyzer. It enables controlled pressurization of reagent bottles to perform accurate calibration and verification procedures.
  • Enables controlled pressurization of reagent bottles for manual calibration and verification.
  • Includes a luer lock syringe, a one-way check valve, and all necessary luer tubing and cap.
  • Designed for use with chlorine analyzers in field or laboratory calibration workflows.
  • Provides quick installation with the included fittings for efficient maintenance.