Learn More
Hach Company CL17 Valve Kit for Calibration Verification - for Hach CL17 chlorine analyzer
Catalog No. NC3717560 Shop All Hach Company Products
Encompass
The CL17 Valve Kit for Calibration Verification (MPN 5449800) is an accessory kit used to perform manual calibration and verification of the CL17 chlorine analyzer. It contains the fluid-routing components required to connect calibration standards and verification reagents, simplifying field and routine maintenance procedures.
  • Includes valves, tubing, and luers to route calibration fluids.
  • Enables manual calibration and verification procedures for accurate measurement.
  • Facilitates quick replacement of worn fluid components to minimize downtime.
  • Designed for straightforward installation and use in field and service settings.
Catalog No. NC3717560 Supplier Hach Company Supplier No. 5449800
The CL17 Valve Kit for Calibration Verification (MPN 5449800) is an accessory kit used to perform manual calibration and verification of the CL17 chlorine analyzer. It contains the fluid-routing components required to connect calibration standards and verification reagents, simplifying field and routine maintenance procedures.
  • Includes valves, tubing, and luers to route calibration fluids.
  • Enables manual calibration and verification procedures for accurate measurement.
  • Facilitates quick replacement of worn fluid components to minimize downtime.
  • Designed for straightforward installation and use in field and service settings.