Learn More
Hach Company CL17sc Cal. Verification Kit
Supplier:  Hach Company 8568200
Encompass
The CL17sc Calibration Verification Kit includes two bottles of deionized water (500 mL) a chlorine standard (20 mL ampule) ampule breaker syringe (50 mL) tubing and fittings The CL17sc calibration curve is set at the factory for performance to specifications User adjustment of the factory calibration curve is only recommended in two instances if required by a regulatory agency for compliance reporting purposes or if a large repair of the analyzer is done
Catalog No. 50-002-07844