Learn More
Hach Company CL17sc Cell Cleaning Kit - maintenance kit for CL17sc colorimeter cell
Catalog No. NC2051612 Shop All Hach Company Products
Encompass
The CL17sc Cell Cleaning Kit is a maintenance accessory designed to clean the colorimetric cell of the CL17sc analyzer. It provides an acid cleaning solution and swabs to remove deposits and help maintain accurate colorimetric measurements.
  • Contains 100 mL of 5.25N sulfuric acid in a dropper bottle.
  • Includes 10 cotton swabs for manual cleaning.
  • Designed for cleaning the colorimetric cell of compatible analyzers.
  • Helps remove deposits that can affect measurement accuracy.
  • Store in a cool, dry, well-ventilated place; protect from moisture.
  • May require alternate shipping methods due to hazardous contents.
Catalog No. NC2051612 Supplier Hach Company Supplier No. 8573100
The CL17sc Cell Cleaning Kit is a maintenance accessory designed to clean the colorimetric cell of the CL17sc analyzer. It provides an acid cleaning solution and swabs to remove deposits and help maintain accurate colorimetric measurements.
  • Contains 100 mL of 5.25N sulfuric acid in a dropper bottle.
  • Includes 10 cotton swabs for manual cleaning.
  • Designed for cleaning the colorimetric cell of compatible analyzers.
  • Helps remove deposits that can affect measurement accuracy.
  • Store in a cool, dry, well-ventilated place; protect from moisture.
  • May require alternate shipping methods due to hazardous contents.