Learn More
Hach Company CL17sc Chlorine Analyzer
Supplier:  Hach Company 8572350
Encompass
CL17sc Chlorine Analyzer with Standpipe Installation Kit ( 10 psi sample pressure to Y-strainer)-and User Manual User provides screws and/or anchors for mounting Hach SC Controller and Reagents are not included-they are sold separately
Catalog No. 50-002-07423