Learn More
Hach Company CL17sc Chlorine Analyzer
Supplier:  Hach Company 8572400
Encompass
CL17sc Chlorine Analyser with Pressure Regulator Installation Kit-and User Manual User provides screws and/or anchors for mounting Hach SC Controller and Reagents are not included-they are sold separately
Catalog No. 50-002-07424