Learn More
Hach Company CL17sc Kit w/SC4500 Ctrl 1 ch
Catalog No. 5000207927 Shop All Hach Company Products
Encompass
CL17sc-Standpipe Kit with SC4500 Controller-1 Channel-24 VDCIncludes SC4500 Controller 24VDC-Prognosys-no plug-single input-controller mounting hardware CL17sc Colorimetric Chlorine Analyzer with Standpipe Installation Kit-w/o Reagents-user manuals
Catalog No. 50-002-07927 Supplier Hach Company Supplier No. LXV525.97KTO061
Add to Cart
Add to Cart
Request a Quote
CL17sc-Standpipe Kit with SC4500 Controller-1 Channel-24 VDCIncludes SC4500 Controller 24VDC-Prognosys-no plug-single input-controller mounting hardware CL17sc Colorimetric Chlorine Analyzer with Standpipe Installation Kit-w/o Reagents-user manuals