Learn More
Medchemexpress LLC Cl2-SN-38 | 1036969-20-6 | 99.2% | 1627.79 | C82H106N12O23 | 5 MG
SDP
Catalog No. 5000203484
Encompass_Preferred
CL2-SN-38 is a cleavable linker-based SN-38 conjugate intended as a payload for antibody-drug conjugate (ADC) research. It enables targeted delivery of the topoisomerase I inhibitor SN-38 by conjugation to antibodies such as the anti-Trop-2 humanized antibody hRS7, and is suitable for in vitro and in vivo cancer research, ADC development, and linker stability studies.
  • Cleavable linker enabling controlled payload release.
  • Contains the topoisomerase I inhibitor SN-38 as the cytotoxic payload.
  • Designed for conjugation to antibodies for ADC development.
  • High purity (>99%) suitable for preclinical research.
  • Available in multiple small-scale quantities for laboratory use.
Catalog No. 50-002-03484 Supplier Medchemexpress LLC Supplier No. HY1263505MG
CL2-SN-38 is a cleavable linker-based SN-38 conjugate intended as a payload for antibody-drug conjugate (ADC) research. It enables targeted delivery of the topoisomerase I inhibitor SN-38 by conjugation to antibodies such as the anti-Trop-2 humanized antibody hRS7, and is suitable for in vitro and in vivo cancer research, ADC development, and linker stability studies.
  • Cleavable linker enabling controlled payload release.
  • Contains the topoisomerase I inhibitor SN-38 as the cytotoxic payload.
  • Designed for conjugation to antibodies for ADC development.
  • High purity (>99%) suitable for preclinical research.
  • Available in multiple small-scale quantities for laboratory use.