Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cl2-sn-38 | 1036969-20-6 | 99.2% | 1627.79 | C82H106N12O23 | 50 MG
SDP

Catalog No. 5000204144
Encompass_Preferred

CL2-SN-38 is a cleavable linker-SN-38 agent-linker conjugate intended for antibody-drug conjugate (ADC) research. It is used to deliver the cytotoxic SN-38 payload to target cells via antibody targeting of cell-surface antigens; intracellular processing releases the active toxin, enabling targeted cancer research applications. Provided data include chemical composition, molecular weight, purity, storage, and solubility guidance.

  • Cleavable linker-SN-38 conjugate for ADC research.
  • Designed to conjugate with Trop-2-targeting antibodies.
  • High purity suitable for research applications.
  • Soluble in DMSO at high concentrations with ultrasonic assistance.
  • Storage guidelines provided for solid and solution forms to maintain stability.
  • Intended for targeted cancer research using ADC methodologies.

Catalog No. 50-002-04144 Supplier Medchemexpress LLC Supplier No. HY12635050MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CL2-SN-38 is a cleavable linker-SN-38 agent-linker conjugate intended for antibody-drug conjugate (ADC) research. It is used to deliver the cytotoxic SN-38 payload to target cells via antibody targeting of cell-surface antigens; intracellular processing releases the active toxin, enabling targeted cancer research applications. Provided data include chemical composition, molecular weight, purity, storage, and solubility guidance.

  • Cleavable linker-SN-38 conjugate for ADC research.
  • Designed to conjugate with Trop-2-targeting antibodies.
  • High purity suitable for research applications.
  • Soluble in DMSO at high concentrations with ultrasonic assistance.
  • Storage guidelines provided for solid and solution forms to maintain stability.
  • Intended for targeted cancer research using ADC methodologies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.