Learn More
Coast CL20R Rechargeable Clamp Light
Catalog No. 17010031
Change view
Click to view available options
Light Source:
LED
Catalog No. Light Source
17-010-031 LED
1 options
Catalog No. 17-010-031 Supplier Coast Supplier No. 30684
Add to Cart
Add to Cart
This Clamp Light takes precision task lighting to an all-new level with multiple positioning options enabled by its high-strength clamp.
  • Outfitted with a Wide Angle Flood Beam to provide a broad sweep of flawless light at 1750 lumens, the clamp on this versatile go-anywhere work light attaches firmly to a wide variety of supports—from ladders to pipes to branches
  • The clamp also doubles as a dynamic stand, or you can use the integrated magnetic base for attachment to metal surfaces
  • Once in place, the light rotates up to 360 degrees and tilts up to 115 degrees for the ultimate in lighting control
  • Engineered with the COAST Dual Power system, the rechargeable CL20R runs on a built-in COAST ZITHION™ battery
  • In addition to the ZITHION™ Rechargeable battery, the CL20R can also be continuously run using standard AC/DC power. To continuously run the Clamp Light, plug the USB-C cable (included) into the Clamp Light and plug the cable into an available power source, then turn the light on. While in a continuous run, the CL20R will operate at a single setting at a reduced level. The clamp light will not charge while in continuous run mode and must be turned off to fully charge. A battery life indicator ensures you know when more power is needed, and the battery also doubles as a power bank for external devices
  • Constructed of top-grade materials, the CL20R is rated IP54 for water resistance
Specifications
Light Source LED
Product Type LED Clip Lamp