Learn More
Medchemexpress LLC CL2A (TFA) | 2616704-22-2 | 99.5% | 1176.24 | C50H79N9O16 | 5 MG
SDP
Catalog No. 5000202270
Encompass_Preferred
CL2A (TFA) is a pH-sensitive, cleavable PEG8- and triazole-containing PABC-peptide-mc linker for use in antibody-drug conjugate construction. The TFA salt form improves handling and stability and is designed to release payloads under acidic conditions to produce a bystander effect.
  • Cleavable linker enabling payload release under acidic conditions.
  • PEG8 spacer and triazole moiety for increased solubility and stability.
  • Designed for conjugation to antibody cysteine residues via disulfide linkage.
  • Available in small research-scale packages for use in ADC development.
  • Stable as a powder when stored sealed at 4°C; shipped at room temperature.
Catalog No. 50-002-02270 Supplier Medchemexpress LLC Supplier No. HY128945A5MG
CL2A (TFA) is a pH-sensitive, cleavable PEG8- and triazole-containing PABC-peptide-mc linker for use in antibody-drug conjugate construction. The TFA salt form improves handling and stability and is designed to release payloads under acidic conditions to produce a bystander effect.
  • Cleavable linker enabling payload release under acidic conditions.
  • PEG8 spacer and triazole moiety for increased solubility and stability.
  • Designed for conjugation to antibody cysteine residues via disulfide linkage.
  • Available in small research-scale packages for use in ADC development.
  • Stable as a powder when stored sealed at 4°C; shipped at room temperature.