Learn More
Cayman Chemical CL2ASN38dIchloroacetIc ac 10mg
SDP
Catalog No. 502947731
Encompass_Preferred
A reactive drug-linker molecule containing SN-38; cytotoxic to various cancer cells when linked to hRS7 as an ADC (IC50s =1.95-23.14 nM); reduces tumor growth in BxPC-3 pancreatic and SK-MES-1 lung cancer mouse xenograft models when linked to hRS7
Catalog No. 50-294-7731 Supplier Cayman Chemical Supplier No. 3394110 MG
A reactive drug-linker molecule containing SN-38; cytotoxic to various cancer cells when linked to hRS7 as an ADC (IC50s =1.95-23.14 nM); reduces tumor growth in BxPC-3 pancreatic and SK-MES-1 lung cancer mouse xenograft models when linked to hRS7