Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cl2e-SN-38 TFA | 99.1% | 1855.95 | C89H117F3N14O26 | 25 MG
SDP

Catalog No. 5000203437
Encompass_Preferred

CL2E-SN38 TFA is a trifluoroacetate salt of an antibody-linked SN-38 conjugate used for antibody-drug conjugate (ADC) research. It acts as a topoisomerase I inhibitor and is supplied as a high-purity solid characterized for in vitro and in vivo formulation.

  • Purity: 99.07% (reported on product page)
  • Form: trifluoroacetate salt
  • Molecular weight: 1855.95
  • Chemical formula: C89H117F3N14O26
  • Solubility: soluble in DMSO (example: 100 mg/mL); in vivo formulations reported at ≥2.5 mg/mL
  • Storage: protect from light; store under nitrogen; in solvent: -80°C for long term
  • Intended use: research reagent for ADC development and topoisomerase I inhibition studies

Catalog No. 50-002-03437 Supplier Medchemexpress LLC Supplier No. HY139909A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CL2E-SN38 TFA is a trifluoroacetate salt of an antibody-linked SN-38 conjugate used for antibody-drug conjugate (ADC) research. It acts as a topoisomerase I inhibitor and is supplied as a high-purity solid characterized for in vitro and in vivo formulation.

  • Purity: 99.07% (reported on product page)
  • Form: trifluoroacetate salt
  • Molecular weight: 1855.95
  • Chemical formula: C89H117F3N14O26
  • Solubility: soluble in DMSO (example: 100 mg/mL); in vivo formulations reported at ≥2.5 mg/mL
  • Storage: protect from light; store under nitrogen; in solvent: -80°C for long term
  • Intended use: research reagent for ADC development and topoisomerase I inhibition studies

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.