Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cl2e-SN-38 TFA | 99.5% | 1855.95 | C89H117F3N14O26 | 1 MG
SDP

Catalog No. 5000203894
Encompass_Preferred

CL2E-SN-38 TFA is an antibody-drug conjugate (ADC) linker-payload provided as the trifluoroacetate (TFA) salt. It incorporates SN-38, the active metabolite of irinotecan, and is intended for research use in ADC development and preclinical studies.

  • Highly releasable linker-SN-38 conjugate suitable for ADC research.
  • Provided as the trifluoroacetate salt to aid handling and formulation.
  • Molecular weight 1855.95 and formula C89H117F3N14O26.
  • High purity (reported ~99.5%).
  • Available in milligram-scale quantities for assay development and conjugation experiments.

Catalog No. 50-002-03894 Supplier Medchemexpress LLC Supplier No. HY139909A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CL2E-SN-38 TFA is an antibody-drug conjugate (ADC) linker-payload provided as the trifluoroacetate (TFA) salt. It incorporates SN-38, the active metabolite of irinotecan, and is intended for research use in ADC development and preclinical studies.

  • Highly releasable linker-SN-38 conjugate suitable for ADC research.
  • Provided as the trifluoroacetate salt to aid handling and formulation.
  • Molecular weight 1855.95 and formula C89H117F3N14O26.
  • High purity (reported ~99.5%).
  • Available in milligram-scale quantities for assay development and conjugation experiments.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.