Learn More
Medchemexpress LLC Cl2e-SN38 TFA | 99.1% | 1855.95 | C89H117F3N14O26 | 5 MG
SDP
Catalog No. 5000204245
Encompass_Preferred
CL2E-SN-38 TFA is an antibody-drug conjugate (ADC) drug-linker consisting of an SN-38 payload attached via a CL2E linker in the trifluoroacetate (TFA) salt form. It is intended for preclinical research into ADCs, where SN-38 functions as a topoisomerase I inhibitor.
  • Highly releasable and structurally stable antibody-SN-38 conjugate.
  • Contains SN-38 payload, a topoisomerase I inhibitor.
  • TFA salt form for improved solubility and handling.
  • Available in small research quantities (5 MG).
  • High purity suitable for preclinical ADC development.
Catalog No. 50-002-04245 Supplier Medchemexpress LLC Supplier No. HY139909A5MG
CL2E-SN-38 TFA is an antibody-drug conjugate (ADC) drug-linker consisting of an SN-38 payload attached via a CL2E linker in the trifluoroacetate (TFA) salt form. It is intended for preclinical research into ADCs, where SN-38 functions as a topoisomerase I inhibitor.
  • Highly releasable and structurally stable antibody-SN-38 conjugate.
  • Contains SN-38 payload, a topoisomerase I inhibitor.
  • TFA salt form for improved solubility and handling.
  • Available in small research quantities (5 MG).
  • High purity suitable for preclinical ADC development.