Learn More
Invivogen CL307, 500 ug
SDP
Catalog No. NC0548959
Encompass
CL307 is a potent and specific Toll-like receptor 7 (TLR7) agonist. It was generated by covalently linking a spermine to the adenine analog CL264, a TLR7 agonist developed by InvivoGen. Coupling with spermine enhances cellular uptake by endocytosis and the subsequent delivery to TLR7.
Catalog No. NC0548959 Supplier Invivogen Supplier No. TLRLC307
CL307 is a potent and specific Toll-like receptor 7 (TLR7) agonist. It was generated by covalently linking a spermine to the adenine analog CL264, a TLR7 agonist developed by InvivoGen. Coupling with spermine enhances cellular uptake by endocytosis and the subsequent delivery to TLR7.