Learn More
Medchemexpress LLC HY-13599 10mg , Cladribine CAS:4291-63-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1359910MG
Encompass_Preferred
HY-13599 10mg Cladribine CAS: 4291-63-8 Cladribine (2-Chloro-2′-deoxyadenosine), a purine nucleoside analog, is an orally active adenosine deaminase inhibitor. Cladribine functions as an inhibitor of DNA synthesis to block the repair of the damaged DNA. Cladribine can inhibit DNA methylation. Cladribine has anti-lymphoma activity. Cladribine can be used for the research of several hematologic malignancies and multiple sclerosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0779