Learn More
New England Biolabs, Inc. ClaI – 1000 units
SDP
Catalog No. 50812131
Encompass
A restriction endonuclease that recognizes the sequence AT^CG_AT.
Catalog No. 50-812-131 Supplier New England Biolabs, Inc. Supplier No. R0197S
A restriction endonuclease that recognizes the sequence AT^CG_AT.