Learn More
New England Biolabs, Inc. ClaI – 5000 units
SDP
Catalog No. 50812130
Encompass
A restriction endonuclease that recognizes the sequence AT^CG_AT.
Catalog No. 50-812-130 Supplier New England Biolabs, Inc. Supplier No. R0197L
A restriction endonuclease that recognizes the sequence AT^CG_AT.