Learn More
Enterprise Technology Solutions Claire Manufacturing Co Cleaner Spray,f/Stainless Steel,Lemon,16oz,12/CT,White/Blue
SDP
Supplier:  Enterprise Technology Solutions CGCCL844
Encompass_Preferred
Water Base Stainless Steel Maintainer cleans, polishes and protects stainless steel at the same time without hard rubbing and polishing. It resists fingerprints, grease and water spatter while helping preserve the factory finish. Use on all types of stainless steel window frames, trims, doors, corridor guards, kickplates, autoclaves, dishwashers, refrigerators, freezers, sterilizers and more in institutions, restaurants, office buildings, hotels/motels, hospitals, washrooms and food-preparation areas. This water-based cleaner is also excellent for chrome. Aerosol spray cleaner comes in a 20 oz. can with a 16 oz. net weight.
Catalog No. 50-008-32676