Learn More
Enterprise Technology Solutions Claire Manufacturing Co Cleaner,Utility,General Purpose,Spray,19 oz,WE
SDP
Catalog No. 5000833113
Encompass_Preferred
Versatile foaming cleaner works on all types of hard surfaces. Use this Gleme Utility Cleaner on walls, doors, appliances, mirrors, windows, counters, furniture or wherever cleaning is needed. Just spray on, allowing the foam to penetrate and break up dirt and grime for easy removal. It cleans plastic surfaces, acrylics, plexiglass, Lexan and polycarbonates. Antistatic, anti-fog cleaner reduces glare and dries in just the right time.
Catalog No. 50-008-33113 Supplier Enterprise Technology Solutions Supplier No. CGCCL862
Versatile foaming cleaner works on all types of hard surfaces. Use this Gleme Utility Cleaner on walls, doors, appliances, mirrors, windows, counters, furniture or wherever cleaning is needed. Just spray on, allowing the foam to penetrate and break up dirt and grime for easy removal. It cleans plastic surfaces, acrylics, plexiglass, Lexan and polycarbonates. Antistatic, anti-fog cleaner reduces glare and dries in just the right time.