Learn More
Enterprise Technology Solutions Claire Manufacturing Co Precision Cleaner,Contact,Anti-Corrosion,13.5 oz
SDP
Catalog No. 5000810635
Encompass_Preferred
P1 Precision Contact Cleaner prevents corrosion by displacing moisture. Formula helps prevent electrical malfunctions caused by water penetration, humidity and condensation. Nonstaining, noncorrosive cleaner is ideal for applications where lower flash point and higher evaporating solvents may be used. Cleaner is safe to use on food-packaging and food-processing equipment and most plastics. Quick-drying formula leaves no residue behind and effectively removes soil and other contaminants from electronic components.
Catalog No. 50-008-10635 Supplier Enterprise Technology Solutions Supplier No. CGCSW293
P1 Precision Contact Cleaner prevents corrosion by displacing moisture. Formula helps prevent electrical malfunctions caused by water penetration, humidity and condensation. Nonstaining, noncorrosive cleaner is ideal for applications where lower flash point and higher evaporating solvents may be used. Cleaner is safe to use on food-packaging and food-processing equipment and most plastics. Quick-drying formula leaves no residue behind and effectively removes soil and other contaminants from electronic components.