Learn More
Enterprise Technology Solutions Claire Manufacturing Co Silicone Spray,f/Food-Processing Equipment,11 oz
SDP
Supplier:  Enterprise Technology Solutions CGCCL948
Encompass_Preferred
Heat Stable Silicone Release Agent is a protective spray to stop food-packaging and food-processing equipment from sticking or binding. It is perfect to use on metal, rubber and plastics as a lubricant, release agent, water-repellent and protective coating. Dry silicone film is nonflammable, colorless, nonstaining, waxless and heat stable. Waterproof, anticorrosive, NSF-registered formula is unaffected by temperatures from negative 40 degrees Fahrenheit up to 200 degrees Fahrenheit. Use on equipment such as ovens, freezers, conveyors, belts and chutes. Spray only on cool ovens, and avoid spraying pilot lights. Proper use of this silicone spray minimizes cleanup time, speeds up equipment disassembly, stops sticking and binding, protects equipment and increases production.
Catalog No. 50-008-33114