Learn More
Enterprise Technology Solutions Claire Manufacturing Co Stain Remover,f/Carpet & Upholstery,Deodorizing,20 oz,BK/MI
SDP
Catalog No. 5000832265
Encompass_Preferred
Sprayways's Instant Carpet Spot Remover uses the latest technology in surfactants, wetting agents and solvents. Powerful, fast-acting formula removes water and oil-based spots and stains from carpets and upholstery. Saturate stains with this high-tech cleaner to dissolve difficult-to-remove materials like butter, crayon, makeup, ink and more. Deodorizing solution will not soften or damage carpet backing.
Catalog No. 50-008-32265 Supplier Enterprise Technology Solutions Supplier No. CGCSW879
Sprayways's Instant Carpet Spot Remover uses the latest technology in surfactants, wetting agents and solvents. Powerful, fast-acting formula removes water and oil-based spots and stains from carpets and upholstery. Saturate stains with this high-tech cleaner to dissolve difficult-to-remove materials like butter, crayon, makeup, ink and more. Deodorizing solution will not soften or damage carpet backing.