Learn More
Enterprise Technology Solutions Claire Manufacturing Co Wipes,f/Stainless Steel,CitrusScent,9-1/2"x12",Tub, 40 Wipes
SDP
Supplier:  Enterprise Technology Solutions CGCCL993
Encompass_Preferred
Stainless steel wipes clean, polish, brighten and remove hard-water stains, smudges, fingerprints and residue while restoring a brilliant shine. Wipes are premoistened with a powerful, citrus-based solution that effectively removes grime but will not harm stainless steel. Formula leaves behind a clear, protective coating to preserve the surface and prevent watermarks and corrosion. Oil-based cleaner wipes are VOC-compliant. They are ideal for use on all stainless steel surfaces such as dishwashers, refrigerators, freezers, elevators, escalators, doors, trims, water fountains, counters, coffee machines and restroom surfaces.
Catalog No. 50-008-33022