Learn More
Chemglass Life Sciences Adapter, Claisen, 19/22 Joint Size, 128mm H x 70mm W
SDP
Catalog No. CG102019
Encompass_Preferred
Adapter, Claisen, 19/22 Joint Size, 128mm H x 70mm W. Outer standard taper joints on both top arms and lower inner joint. All three joints are the same size.
Catalog No. CG102019 Supplier Chemglass Life Sciences Supplier No. CG102019
Adapter, Claisen, 19/22 Joint Size, 128mm H x 70mm W. Outer standard taper joints on both top arms and lower inner joint. All three joints are the same size.