Learn More
Chemglass Life Sciences Adapter, Claisen, 24/40 Joint, Modified
SDP
Catalog No. CG107401
Encompass_Preferred
Adapter, Claisen, 24/40 Joint, Modified. Claisen style adapter with an additional standard taper outer joint at a 45º angle to the vertical outer joint.
Catalog No. CG107401 Supplier Chemglass Life Sciences Supplier No. CG107401
Adapter, Claisen, 24/40 Joint, Modified. Claisen style adapter with an additional standard taper outer joint at a 45º angle to the vertical outer joint.