Learn More
Ace Glass, Inc. CLAMP 19MM CLAMP 19MM
SDP
Supplier:  Ace Glass, Inc. 781852
Encompass_Preferred
19mm split-ring clamp with hardware, for the ACE-Burlitch system. The ACE-Burlitch inert atmosphere system is designed for maximum convenience and versatility in the handling of air-sensitive compounds in conjunction with ACE no-air labware, or suitiably adapted conventional glassware.
Catalog No. 50-975-474