Learn More
Hach Company Clamp, Two-Prong Extension
Supplier:  Hach Company 2114500
Encompass
Flat, vinyl-covered jaws hold firmly at any angle. One jaw is fixed; the other opens against a stainless steel compression spring. Clamps are nickel-plated zinc with seamless brass extension tube, compressing screw, and pivot pin. Jaw capacity: 3.8 cm (1.5"). Total length: 20 cm (7.8").
Catalog No. 50-199-5433