Learn More
Chemglass Life Sciences Chemglass PTFE Clamp for 24/40 Standard Taper Joint, Green
SDP
Catalog No. NC2457425
Encompass_Preferred
Chemglass PTFE clamp designed to secure 24/40 standard taper joints in laboratory glassware assemblies. The color-coded green clamp is molded from chemically resistant PTFE with an internal steel core for strength and can withstand temperatures up to 250°C.
  • Molded PTFE construction with steel core for strength.
  • Designed to fit 24/40 standard taper joints.
  • Color-coded green for quick identification.
  • Chemically resistant and inert.
  • Withstands temperatures up to 250°C.
Catalog No. NC2457425 Supplier Chemglass Life Sciences Supplier No. CG145P24
Chemglass PTFE clamp designed to secure 24/40 standard taper joints in laboratory glassware assemblies. The color-coded green clamp is molded from chemically resistant PTFE with an internal steel core for strength and can withstand temperatures up to 250°C.
  • Molded PTFE construction with steel core for strength.
  • Designed to fit 24/40 standard taper joints.
  • Color-coded green for quick identification.
  • Chemically resistant and inert.
  • Withstands temperatures up to 250°C.