Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, Polypropylene, Straight, Sanitary Clamp to Hosebarb Adapter with TPE Fused Gasket, Gamma Irradiated, 3/4"" Mini Clamp x 1/4"" ID; 10/pk
Catalog No. NC1958304
Encompass
Sanitary fitting with built-in gasket makes sanitary installation easier than ever. Presterilized adapters cut down on lead time and are available for quick shipment. Sanitary gasket combined with a sanitary fitting adds another layer of safety to installation. Prevent contamination with zero user contact with the gasket to ensure sterility.
Catalog No. NC1958304 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3120116
Sanitary fitting with built-in gasket makes sanitary installation easier than ever. Presterilized adapters cut down on lead time and are available for quick shipment. Sanitary gasket combined with a sanitary fitting adds another layer of safety to installation. Prevent contamination with zero user contact with the gasket to ensure sterility.