Learn More
Hach Company Clamp, Three-Prong Universal
Supplier:  Hach Company 42200
Encompass
Vinyl-covered jaws operate independently of solid aluminum extension rod, 1.1-cm (0.43") diameter. Jaw capacity: 8.9 cm (3.5"). Brass pivot pin. Total length: 27.9 cm (10.9").
Catalog No. 50-199-4279