Learn More
Ace Glass, Inc. CLAMP 32MM CLAMP 32MM
SDP
Catalog No. 50975475
Encompass_Preferred
32mm split-ring clamp with hardware, for the ACE-Burlitch system. The ACE-Burlitch inert atmosphere system is designed for maximum convenience and versatility in the handling of air-sensitive compounds in conjunction with ACE no-air labware, or suitiably adapted conventional glassware.
Catalog No. 50-975-475 Supplier Ace Glass, Inc. Supplier No. 781854
32mm split-ring clamp with hardware, for the ACE-Burlitch system. The ACE-Burlitch inert atmosphere system is designed for maximum convenience and versatility in the handling of air-sensitive compounds in conjunction with ACE no-air labware, or suitiably adapted conventional glassware.