Learn More
Chemglass Life Sciences Clamp, Two-Prong Extension, Large, Single Adjust, 9.02in OAL
SDP
Catalog No. CG920202
Encompass_Preferred
Clamp, Two-Prong Extension, Large, Single Adjust, 9in OAL. Nickel-plated zinc alloy clamp has high tensile strength and corrosion resistance. Fully enclosed stainless steel springs provide smooth operation. Two-prong clamps are available in single and dual adjustment. The medium clamp has a 1 1/2¿ (38mm) maximum grip size with a 1/2¿ (13mm) O.D. extension arm. The large clamp has a 2 1/2¿ (64mm) maximum grip size with a 5/8¿ (16mm) O.D. extension arm. Complete with removable vinyl and fiberglass sleeve covers.
Catalog No. CG920202 Supplier Chemglass Life Sciences Supplier No. CG920202
Clamp, Two-Prong Extension, Large, Single Adjust, 9in OAL. Nickel-plated zinc alloy clamp has high tensile strength and corrosion resistance. Fully enclosed stainless steel springs provide smooth operation. Two-prong clamps are available in single and dual adjustment. The medium clamp has a 1 1/2¿ (38mm) maximum grip size with a 1/2¿ (13mm) O.D. extension arm. The large clamp has a 2 1/2¿ (64mm) maximum grip size with a 5/8¿ (16mm) O.D. extension arm. Complete with removable vinyl and fiberglass sleeve covers.