Learn More
Andwin Scientific Talon Three-Prong Extension Clamps, Medim, Zinc, 127mm Extension size
SDP
Catalog No. NC9610241
Encompass_Preferred
With a large grip adjustment range, the Talon Three-Prong Extension Clamps can safely hold every type of glassware and apparatus. This product features round extension arms that allow the clamps to be rotated 360. It is constructed with Zinc materi
Catalog No. NC9610241 Supplier Andwin Scientific Supplier No. 21570302
With a large grip adjustment range, the Talon Three-Prong Extension Clamps can safely hold every type of glassware and apparatus. This product features round extension arms that allow the clamps to be rotated 360. It is constructed with Zinc materi