Learn More
Ace Glass, Inc. Clamp, 60mm ID, bolt latch closure, Viton; inner cushions, 5/8in OD x 12in length rod
SDP
Catalog No. 50978734
Encompass
60mm rod-mounted bolt latch clamp secures the top of 100mL to 200L reaction flasks to the stand.
Catalog No. 50-978-734 Supplier Ace Glass, Inc. Supplier No. 644202
60mm rod-mounted bolt latch clamp secures the top of 100mL to 200L reaction flasks to the stand.