Learn More
Med Vet International Miltex Aortic Aneurysm Clamp, Slightly Curved Jaws, 12.25in, German, 24-1408
Catalog No. 502830670
Encompass_Preferred
Clamps that are ideally used in stopping blood flow at the neck of sacciform aneurysms. 12.25in. Made with high quality German steel.
Catalog No. 50-283-0670 Supplier Med Vet International Supplier No. MIL241408
Clamps that are ideally used in stopping blood flow at the neck of sacciform aneurysms. 12.25in. Made with high quality German steel.