Learn More
Chemglass Life Sciences Chain Clamp, 12" Arm
SDP
Catalog No. 501311238
Encompass_Preferred
Chain Clamp, 12" Arm. Similar to the CG-9220-01 except has a 12" long arm. Clamp can accommodate up to 6-1/2¿ (170mm) O.D. respectively. Quick slip-on chain connection has large, easy-to-turn adjusting knob. Grip is vinyl coated.
Catalog No. 50-131-1238 Supplier Chemglass Life Sciences Supplier No. CG9220E01
Chain Clamp, 12" Arm. Similar to the CG-9220-01 except has a 12" long arm. Clamp can accommodate up to 6-1/2¿ (170mm) O.D. respectively. Quick slip-on chain connection has large, easy-to-turn adjusting knob. Grip is vinyl coated.