Learn More
Ace Glass, Inc. Clamp, column, 64mm min to 140mm max, 338mm overall length with arm
SDP
Catalog No. 50977186
Encompass
Column clamp IDeal for use with cylindrical glassware having an outside diameter of 64mm to 140mm. A large, flat thumbscrew/worm-drive permits maximum tightening or removal in minimum time. The clamp is constructed of corrosion-resistant stainless steel, with an 11mm D x 211mm L rod extending from the clamp for easy mounting. 338mm overall length, including arm.
Catalog No. 50-977-186 Supplier Ace Glass, Inc. Supplier No. 1107806
Column clamp IDeal for use with cylindrical glassware having an outside diameter of 64mm to 140mm. A large, flat thumbscrew/worm-drive permits maximum tightening or removal in minimum time. The clamp is constructed of corrosion-resistant stainless steel, with an 11mm D x 211mm L rod extending from the clamp for easy mounting. 338mm overall length, including arm.