Learn More
Ace Glass, Inc. Clamp, column, 91mm min to 114mm max, 315mm overall length with arm
SDP
Catalog No. 50977185
Encompass
Column clamp IDeal for use with cylindrical glassware having an outside diameter of 91mm to 114mm. A large, flat thumbscrew/worm-drive permits maximum tightening or removal in minimum time. The clamp is constructed of corrosion-resistant stainless steel, with an 11mm D x 211mm L rod extending from the clamp for easy mounting. 315mm overall length, including arm.
Catalog No. 50-977-185 Supplier Ace Glass, Inc. Supplier No. 1107804
Column clamp IDeal for use with cylindrical glassware having an outside diameter of 91mm to 114mm. A large, flat thumbscrew/worm-drive permits maximum tightening or removal in minimum time. The clamp is constructed of corrosion-resistant stainless steel, with an 11mm D x 211mm L rod extending from the clamp for easy mounting. 315mm overall length, including arm.