Learn More
Med Vet International Miltex Debakey Bulldog Clamp, Angled, Cross-Action, 3in, German, 24-650
Catalog No. 502829539
Encompass_Preferred
Bulldog clamps are used for stopping blood flow during cardiothoracic procedures. Includes a angled jaw and cross action handles for the pinnacle of precision and control. 3in. 0.75in jaw length. Made with long-lasting, high-quality German steel.
Catalog No. 50-282-9539 Supplier Med Vet International Supplier No. MIL24650
Bulldog clamps are used for stopping blood flow during cardiothoracic procedures. Includes a angled jaw and cross action handles for the pinnacle of precision and control. 3in. 0.75in jaw length. Made with long-lasting, high-quality German steel.