Learn More
Med Vet International Miltex Debakey Bulldog Clamp, Straight, Cross-Action, 4.12in, German, 24-604
Catalog No. 502829538
Encompass_Preferred
Bulldog clamps are used for stopping blood flow during cardiothoracic procedures. Includes a straight jaw and cross action handles for the pinnacle of precision and control. 4.12in. Made with long-lasting, high-quality German steel.
Catalog No. 50-282-9538 Supplier Med Vet International Supplier No. MIL24604
Bulldog clamps are used for stopping blood flow during cardiothoracic procedures. Includes a straight jaw and cross action handles for the pinnacle of precision and control. 4.12in. Made with long-lasting, high-quality German steel.