Learn More
Med Vet International Miltex Debakey Patent Ductus & Peripheral Vascular Clamp, Straight, 7.75in, German, 24-1150
Catalog No. 502830659
Encompass_Preferred
Multipurpose clamp designed for use in a number of different cardiothoracic surgical procedures. Well suited for aortic and pulmonary wall clamping during suturing and division of the ductus arteriosus in patients with patent ductus arteriosus. 7.75in. 32mm jaw. Made with high quality German steel.
Catalog No. 50-283-0659 Supplier Med Vet International Supplier No. MIL241150
Multipurpose clamp designed for use in a number of different cardiothoracic surgical procedures. Well suited for aortic and pulmonary wall clamping during suturing and division of the ductus arteriosus in patients with patent ductus arteriosus. 7.75in. 32mm jaw. Made with high quality German steel.