Learn More
Med Vet International Miltex Debakey-Satinsky Vena Clamp, Small, 9.5in, German, 24-2100
Catalog No. 502829455
Encompass_Preferred
Highly versatile cardiac clamp that can be used for a wide variety of scenarios that require clamping of a vessel for partial or total occlusion of a vessel but this is designed for use in clamping the vena cava. The slightly angled u-shaped jaw provides adequate grip on the vessel without causing injury to the vessel walls. 9.5in. Small. Made with high quality German steel.
Catalog No. 50-282-9455 Supplier Med Vet International Supplier No. MIL242100
Highly versatile cardiac clamp that can be used for a wide variety of scenarios that require clamping of a vessel for partial or total occlusion of a vessel but this is designed for use in clamping the vena cava. The slightly angled u-shaped jaw provides adequate grip on the vessel without causing injury to the vessel walls. 9.5in. Small. Made with high quality German steel.