Learn More
Med Vet International Miltex Doyen Intestinal Clamp, Curved, Longitudinal Serrations, 9in, German, Mh16-162
Supplier:  Med Vet International MILMH16162
Encompass_Preferred
Designed for a variety of intestinal procedures. Blades with longitudinal serrations. Equipped with a finger ring handle, a ratchet lock, and curved flexible blades. MeisterHand build quality provides exceptional craftsmanship. Made with high-quality German steel. 9in.
Catalog No. 50-283-1628