Learn More
Med Vet International Miltex Doyen Intestinal Clamp, Straight, Diagonal Serrations, 7in, German, 16-108
Catalog No. 502831271
Encompass_Preferred
Designed for a variety of intestinal procedures. Equipped with a finger ring handle, a ratchet lock, and straight flexible blades. Made with high-quality German steel. 7in.
Catalog No. 50-283-1271 Supplier Med Vet International Supplier No. MIL16108
Designed for a variety of intestinal procedures. Equipped with a finger ring handle, a ratchet lock, and straight flexible blades. Made with high-quality German steel. 7in.