Learn More
Wilmad Labglass CLAMP EXT DUAL LRG 3 1/2IN
Catalog No. 50801134
Encompass
Clamp, Extension, Three Prong, Dual Adjustment. Maximum Grip Size: 3- 1/2 in. (89mm); Maximum Load: 20kg; Overall Length: 10- 1/2 in. (27cm), Clamp Extension: 6 in. (15cm)
Catalog No. 50-801-134 Supplier Wilmad Labglass Supplier No. LG4637110
Clamp, Extension, Three Prong, Dual Adjustment. Maximum Grip Size: 3- 1/2 in. (89mm); Maximum Load: 20kg; Overall Length: 10- 1/2 in. (27cm), Clamp Extension: 6 in. (15cm)